missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD45RO (T-Cell Marker) Ab-1 Mouse Monoclonal Antibody, Epredia™
Obtain accurate, reproducible results in immunohistochemistry and flow cytometry experiments with Epredia™ CD45RO (T-Cell Marker) Ab-1, Mouse Monoclonal Antibody.
£99.30 - £250.40
Specifications
Antigen | CD45RO (T-Cell Marker) Ab-1 |
---|---|
Clone | UCHL-1 |
Applications | Flow Cytometry, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Conjugate | Unconjugated |
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
12699056
|
Epredia
MS-112-P1 |
Concentrated | 500 μL |
£190.00
500µL |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12609066
|
Epredia
MS-112-P1ABX |
Concentrated | 100 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12619066
|
Epredia
MS-112-PABX |
Concentrated | 200 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12689056
|
Epredia
MS-112-P0 |
Concentrated | 100 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12679056
|
Epredia
MS-112-P |
Concentrated | 1 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12649066
|
Epredia
MS-112-R7 |
Prediluted | 7 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12659066
|
Epredia
MS-112-RQ |
Ready-to-Use | 12 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
CD45RO (an isoform of leukocyte common antigen) is expressed on most thymocytes, monocytes, macrophages and granulocytes. It is not expressed on normal B lymphocytes. Intrathymic T cell maturation progresses from CD45RA-/CD45RO+ to CD45RA+/CD45RO-; while peripheral T cell maturation progresses from CD45RA+/CD45RO- to CD45RA-/CD45RO+.Host Species: Mouse
Clone: UCHL-1
Isotype: IgG2a/κ
Species Reactivity: Human, Rhesus Monkey and Chimpanzee. Does not react with rat. Others not known
Epitope: Not determined
Immunogen: Interleukin-2-dependent human T lymphocytes
Molecular Weight: 180 kDa
Positive Control: Tonsil or lymph node
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Flow Cytometry
Specifications
CD45RO (T-Cell Marker) Ab-1 | |
Flow Cytometry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
IVD | |
Interleukin-2-dependent human T lymphocytes |
UCHL-1 | |
Monoclonal | |
Mouse | |
Oncogenesis | |
Primary |