missing translation for 'onlineSavingsMsg'
Learn More

CD45RO (T-Cell Marker) Ab-1 Mouse Monoclonal Antibody, Epredia™

Obtain accurate, reproducible results in immunohistochemistry and flow cytometry experiments with Epredia™ CD45RO (T-Cell Marker) Ab-1, Mouse Monoclonal Antibody.

Brand:  Epredia MS-112-P1ABX

Code : LD

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 12609066

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

CD45RO (an isoform of leukocyte common antigen) is expressed on most thymocytes, monocytes, macrophages and granulocytes. It is not expressed on normal B lymphocytes. Intrathymic T cell maturation progresses from CD45RA-/CD45RO+ to CD45RA+/CD45RO-; while peripheral T cell maturation progresses from CD45RA+/CD45RO- to CD45RA-/CD45RO+.

Host Species: Mouse

Clone: UCHL-1

Isotype: IgG2a/κ

Species Reactivity: Human, Rhesus Monkey and Chimpanzee. Does not react with rat. Others not known

Epitope: Not determined

Immunogen: Interleukin-2-dependent human T lymphocytes

Molecular Weight: 180 kDa

Positive Control: Tonsil or lymph node

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Flow Cytometry
Specifications

Specifications

CD45RO (T-Cell Marker) Ab-1
Monoclonal
1mg/mL
Mouse
100 μL
Oncogenesis
Human, Non-human Primate
IgG2a κ
Flow Cytometry, Immunohistochemistry (Paraffin)
UCHL-1
Unconjugated
Interleukin-2-dependent human T lymphocytes
IVD
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers