missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IBA Lifesciences CD4 Isolation Kit for FABian&trade, Mouse
For automated positive cell separation. The Isolation Kit for FABian™ provides label-free target cells without the use of high affinity antibodies and magnetic beads
Brand: IBA Lifesciences 6-6501-010
Additional Details : Weight : 0.90000kg
Description
Specifications
Isolation of label-free cells | |
2°C to 8°C | |
3x107 target cells/column |
Non-magnetic Cell Isolation Kit | |
Mouse Spleen | |
10 Kits |