missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IBA Lifesciences CD4 Isolation Kit for FABian&trade, Mouse
For automated positive cell separation. The Isolation Kit for FABian™ provides label-free target cells without the use of high affinity antibodies and magnetic beads
£84.14 - £664.86
Specifications
Description | Isolation of label-free cells |
---|---|
Product Type | Non-magnetic Cell Isolation Kit |
Content And Storage | 2°C to 8°C |
Donor Source | Mouse Spleen |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16242931
|
IBA Lifesciences
6-6501-001 |
1 Kit |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16252931
|
IBA Lifesciences
6-6501-010 |
10 Kits |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Specifications
Isolation of label-free cells | |
Non-magnetic Cell Isolation Kit | |
2°C to 8°C | |
Mouse Spleen |