Learn More
IBA Lifesciences CD4 Isolation Kit for FABian&trade, Mouse
For automated positive cell separation. The Isolation Kit for FABian™ provides label-free target cells without the use of high affinity antibodies and magnetic beads
Brand:  IBA Lifesciences 6-6501-001
Additional Details : Weight : 0.09000kg
 View more versions of this product
Product Code. 16242931
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • The kit is used for the selection of CD4+ T cells from mouse spleen in conjunction with the FABian&trade system.
  • It includes CD4 Fab-Strep (mouse), the Cell Isolation Column, Cell Isolation Buffer, Biotin for Cell Elution and tubes and a luer lock adapter with the syringe.
  • Each kit is sufficient for one FABian&trade run to isolate up to 3x10^7 target cells.
  • Specifications
    Specifications
    Isolation of label-free cells
    2°C to 8°C
    3x107 target cells
    Non-magnetic Cell Isolation Kit
    Mouse Spleen
    1 Kit