missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ CD20 Rabbit Monoclonal Antibody supernatant
CD20, Rabbit Monoclonal Ab is part of a comprehensive ab portfolio. Studies show rabbit monoclonal antibodies are more sensitive and specific than other available markers.
£118.32 - £540.60
Specifications
Clone | EP459Y |
---|---|
Classification | Monoclonal |
Conjugate | Unconjugated |
Form | Concentrated |
Host Species | Rabbit |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15230685
|
Epredia™
RM-2115-S0 |
100 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15200685
|
Epredia™
RM-2115-R7 |
7 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15210685
|
Epredia™
RM-2115-RQ |
12 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15220685
|
Epredia™
RM-2115-S |
501 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
CD20 is a non-Ig differentiation antigen of B cells and the expression of CD20 is restricted to normal and neoplastic B cells, being absent from all other leukocytes and tissues. It acts as calcium channel involved in B cell activation and cell cycle progression.Host Species: Rabbit
Clone: EP459Y
Isotype: Rabbit IgG
Species Reactivity: Human. Others not tested.
Immunogen: A synthetic peptide corresponding the residues in the C-term of human CD20
Molecular Weight: 35kDa
Positive Control: Tonsil
Cellular Localization: Cell Membrane/cytoplasm
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
EP459Y | |
Unconjugated | |
Rabbit | |
Human | |
IgG |
Monoclonal | |
Concentrated | |
IVD | |
A synthetic peptide corresponding the residues in the C-term of human CD20 | |
Primary |