missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ CD20 Rabbit Monoclonal Antibody supernatant

CD20, Rabbit Monoclonal Ab is part of a comprehensive ab portfolio. Studies show rabbit monoclonal antibodies are more sensitive and specific than other available markers.

Brand:  Epredia™ RM-2115-S

Code : NEW

Additional Details : Weight : 0.00250kg

 View more versions of this product

Product Code. 15220685

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

CD20 is a non-Ig differentiation antigen of B cells and the expression of CD20 is restricted to normal and neoplastic B cells, being absent from all other leukocytes and tissues. It acts as calcium channel involved in B cell activation and cell cycle progression.

Host Species: Rabbit

Clone: EP459Y

Isotype: Rabbit IgG

Species Reactivity: Human. Others not tested.

Immunogen: A synthetic peptide corresponding the residues in the C-term of human CD20

Molecular Weight: 35kDa

Positive Control: Tonsil

Cellular Localization: Cell Membrane/cytoplasm

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
Specifications

Specifications

Monoclonal
Unconjugated
A synthetic peptide corresponding the residues in the C-term of human CD20
IVD
Human
IgG
EP459Y
Rabbit
501 mL
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers