Learn More
Epredia™ CD20 Rabbit Monoclonal Antibody supernatant
CD20, Rabbit Monoclonal Ab is part of a comprehensive ab portfolio. Studies show rabbit monoclonal antibodies are more sensitive and specific than other available markers.
Brand: Epredia™ RM-2115-RQ
Code : NEW
Additional Details : Weight : 0.02000kg
Description
CD20 is a non-Ig differentiation antigen of B cells and the expression of CD20 is restricted to normal and neoplastic B cells, being absent from all other leukocytes and tissues. It acts as calcium channel involved in B cell activation and cell cycle progression.Host Species: Rabbit
Clone: EP459Y
Isotype: Rabbit IgG
Species Reactivity: Human. Others not tested.
Immunogen: A synthetic peptide corresponding the residues in the C-term of human CD20
Molecular Weight: 35kDa
Positive Control: Tonsil
Cellular Localization: Cell Membrane/cytoplasm
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
Immunohistochemistry (Paraffin) | |
EP459Y | |
Rabbit | |
12 mL | |
Primary | |
Concentrated |
Monoclonal | |
Unconjugated | |
A synthetic peptide corresponding the residues in the C-term of human CD20 | |
IVD | |
Human | |
IgG |