missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD20 Mouse, anti-Human, CoraLite Plus 750, Clone: 4A7G3, Proteintech
Mouse Monoclonal Antibody
£159.65 - £416.33
Specifications
| Antigen | CD20 |
|---|---|
| Clone | 4A7G3 |
| Concentration | 1000 μg/mL |
| Applications | Flow Cytometry (Intracellular Staining) |
| Conjugate | CoraLite Plus 750 |
Description
The CD20 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CD20. This antibody recognizes human antigen. The CD20 antibody has been validated for the following applications: FC (Intra) analysis. This antibody is coralite™ plus 750.
Specifications
| CD20 | |
| 1000 μg/mL | |
| CoraLite Plus 750 | |
| Mouse | |
| 931 | |
| IgG2b | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
| 4A7G3 | |
| Flow Cytometry (Intracellular Staining) | |
| Liquid | |
| Human | |
| B lymphocyte antigen CD20, B1, Bp35, CD20, LEU 16, MS4A1, MS4A1,CD20, MS4A2, S7 | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title