Learn More
CD20 Mouse, anti-Human, CoraLite Plus 750, Clone: 4A7G3, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL750-60271-20UL
Description
The CD20 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CD20. This antibody recognizes human antigen. The CD20 antibody has been validated for the following applications: FC (Intra) analysis. This antibody is coralite™ plus 750.
Specifications
| CD20 | |
| 4A7G3 | |
| CoraLite Plus 750 | |
| Mouse | |
| Primary | |
| Human | |
| 155 kDa | |
| IgG2b |
| Flow Cytometry (Intracellular Staining) | |
| 1000 μg/mL | |
| B lymphocyte antigen CD20, B1, Bp35, CD20, LEU 16, MS4A1, MS4A1,CD20, MS4A2, S7 | |
| 20 μL | |
| 931 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |