Learn More
CD20 Mouse, anti-Human, CoraLite Plus 750, Clone: 4A7G3, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech CL750-60271-100UL
 View more versions of this product
Product Code. 18086865
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
The CD20 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CD20. This antibody recognizes human antigen. The CD20 antibody has been validated for the following applications: FC (Intra) analysis. This antibody is coralite™ plus 750.
Specifications
Specifications
CD20
4A7G3
CoraLite Plus 750
Mouse
Primary
Human
155 kDa
IgG2b
Flow Cytometry (Intracellular Staining)
1000 μg/mL
B lymphocyte antigen CD20, B1, Bp35, CD20, LEU 16, MS4A1, MS4A1,CD20, MS4A2, S7
100 μL
931
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.