Learn More
CD16/CD32 Rat anti-Mouse, PE, Clone: 2.4G2, BD
Rat Monoclonal Antibody
£83.80 - £300.37
Specifications
| Antigen | CD16/CD32 |
|---|---|
| Clone | 2.4G2 |
| Concentration | 0.2mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15821279
|
BD Biosciences
561727 |
50 μg |
£87.75 £83.80 / Each Save £3.95 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15807038
|
BD Biosciences
553145 |
0.2 mg |
£313.00 £300.37 / Each Save £12.63 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The 2.4G2 antibody reacts specifically with a common nonpolymorphic epitope on the extracellular domains of the mouse FcγIII and FcγII receptors. It has also been reported to bind the FcγI receptor (CD64) via its Fc domain. 2.4G2 mAb blocks non-antigen-specific binding of immunoglobulins to the FcγIII and FcγII, and possibly FcγI, receptors . CD16 and/or CD32 are expressed on natural killer cells, monocytes, macrophages, dendritic cells (at low levels), Kupffer cells, granulocytes, mast cells, B lymphocytes, immature thymocytes, and some activated mature T lymphocytes.
Specifications
| CD16/CD32 | |
| 0.2mg/mL | |
| Monoclonal | |
| Rat | |
| Mouse | |
| Mouse BALB/c Macrophage J774 | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| 2.4G2 | |
| Flow Cytometry | |
| PE | |
| RUO | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| IgG2b κ | |
| Affinity Purified | |
| FcγRIII/FcγRII; Fcgr3/Fcgr2 |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.