missing translation for 'onlineSavingsMsg'
Learn More

CD16/CD32 Rat anti-Mouse, PE, Clone: 2.4G2, BD

Rat Monoclonal Antibody

Brand:  BD Biosciences 561727

 View more versions of this product

Product Code. 15821279

  • ONLINE EXCLUSIVE PRICE
    £87.75
    £83.80 / Each
    Save £3.95
    4% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The 2.4G2 antibody reacts specifically with a common nonpolymorphic epitope on the extracellular domains of the mouse FcγIII and FcγII receptors. It has also been reported to bind the FcγI receptor (CD64) via its Fc domain. 2.4G2 mAb blocks non-antigen-specific binding of immunoglobulins to the FcγIII and FcγII, and possibly FcγI, receptors . CD16 and/or CD32 are expressed on natural killer cells, monocytes, macrophages, dendritic cells (at low levels), Kupffer cells, granulocytes, mast cells, B lymphocytes, immature thymocytes, and some activated mature T lymphocytes.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD16/CD32
Monoclonal
0.2mg/mL
FcγRIII/FcγRII; Fcgr3/Fcgr2
Rat
Affinity Purified
RUO
Mouse
IgG2b κ
Flow Cytometry
2.4G2
PE
Aqueous buffered solution containing ≤0.09% sodium azide.
Mouse BALB/c Macrophage J774
50 μg
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.