Learn More
CD16/CD32 Rat anti-Mouse, PE, Clone: 2.4G2, BD
Rat Monoclonal Antibody
Brand: BD Biosciences 553145
Description
The 2.4G2 antibody reacts specifically with a common nonpolymorphic epitope on the extracellular domains of the mouse FcγIII and FcγII receptors. It has also been reported to bind the FcγI receptor (CD64) via its Fc domain. 2.4G2 mAb blocks non-antigen-specific binding of immunoglobulins to the FcγIII and FcγII, and possibly FcγI, receptors . CD16 and/or CD32 are expressed on natural killer cells, monocytes, macrophages, dendritic cells (at low levels), Kupffer cells, granulocytes, mast cells, B lymphocytes, immature thymocytes, and some activated mature T lymphocytes.
Specifications
| CD16/CD32 | |
| Monoclonal | |
| 0.2mg/mL | |
| FcγRIII/FcγRII; Fcgr3/Fcgr2 | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| 2.4G2 | |
| PE | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse BALB/c Macrophage J774 | |
| 0.2 mg | |
| Immunology | |
| Mouse | |
| IgG2b κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.