missing translation for 'onlineSavingsMsg'
Learn More
Learn More
β-Catenin Rabbit Polyclonal Antibody, Epredia™
Stain formalin-fixed, paraffin-embedded tissues with Epredia™ Catenin, β, Rabbit Polyclonal Antibody.
£121.38 - £617.10
Specifications
Antigen | Catenin, beta |
---|---|
Applications | Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
12663377
|
Epredia
RB-9035-P1 |
Concentrated | 500 μL |
£441.00
500µL |
Estimated Shipment: 28-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12653377
|
Epredia
RB-9035-P0 |
Concentrated | 100 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12643377
|
Epredia
RB-9035-P |
Concentrated | 1 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12693377
|
Epredia
RB-9035-R7 |
Prediluted | 7 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
The catenins (α, Β and γ) are ubiquitously expressed cytoplasmic proteins associated with E-cadherin at cellular junctions. Β-catenin also binds to N-cadherin and co-immunoprecipitates with APC. Cadherin/catenin complexes are linked to the cytoskeleton via a direct association between α-actinin and α-catenin. Increased tyrosine phosphorylation can disrupt catenin-cadherin complexes, influencing cellular adhesion.Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human and Mouse. Others not tested.
Epitope: C-terminal
Immunogen: A synthetic peptide from the C-terminus of the β-catenin protein
Molecular Weight: 92kDa
Positive Control: MCF-7 cells, Breast CA
Cellular Localization: Cytoplasmic and cell membrane
This antibody is excellent for staining of formalin-fixed, paraffin-embedded tissues.
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunoprecipitation
Specifications
Catenin, beta | |
Polyclonal | |
Rabbit | |
Transcription Regulation | |
A synthetic peptide from the C-terminus of the beta-catenin protein | |
Primary |
Immunoprecipitation, Western Blot | |
Unconjugated | |
IVD | |
Human | |
IgG |