Learn More
β-Catenin Rabbit Polyclonal Antibody, Epredia™
Stain formalin-fixed, paraffin-embedded tissues with Epredia™ Catenin, β, Rabbit Polyclonal Antibody.
Brand: Epredia RB-9035-P
Code : LD
Additional Details : Weight : 0.00250kg
Description
The catenins (α, Β and γ) are ubiquitously expressed cytoplasmic proteins associated with E-cadherin at cellular junctions. Β-catenin also binds to N-cadherin and co-immunoprecipitates with APC. Cadherin/catenin complexes are linked to the cytoskeleton via a direct association between α-actinin and α-catenin. Increased tyrosine phosphorylation can disrupt catenin-cadherin complexes, influencing cellular adhesion.Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human and Mouse. Others not tested.
Epitope: C-terminal
Immunogen: A synthetic peptide from the C-terminus of the β-catenin protein
Molecular Weight: 92kDa
Positive Control: MCF-7 cells, Breast CA
Cellular Localization: Cytoplasmic and cell membrane
This antibody is excellent for staining of formalin-fixed, paraffin-embedded tissues.
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunoprecipitation
Specifications
Catenin, beta | |
Polyclonal | |
200µg/mL | |
Rabbit | |
1 mL | |
Transcription Regulation | |
Human | |
IgG |
Immunoprecipitation, Western Blot | |
Polyclonal | |
Unconjugated | |
A synthetic peptide from the C-terminus of the beta-catenin protein | |
IVD | |
Primary | |
Concentrated |