missing translation for 'onlineSavingsMsg'
Learn More

CAPRIN1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

£135.00 - £349.00

Specifications

Antigen CAPRIN1
Concentration 0.21 mg/mL
Applications Western Blot, Immunofluorescence, Immunoprecipitation, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16814144
View Documents
Proteintech
15112-1-AP-20UL
20 μL
£135.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16804144
View Documents
Proteintech
15112-1-AP-150UL
150 μL
£349.00
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

GPIP137 may regulate the transport and translation of mRNAs of proteins involved in synaptic plasticity in neurons and cell proliferation and migration in multiple cell types. It is expressed in the cytoplasm.
Specifications

Specifications

CAPRIN1
Western Blot, Immunofluorescence, Immunoprecipitation, Immunocytochemistry, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Rat, Human, Mouse
Q14444, Q5M9G3, Q60865
362173, 4076, 53872
CAPRIN1 Fusion Protein Ag7099
Primary
-20°C
CAPRIN1
0.21 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
caprin 1, CAPRIN1, GPI anchored protein p137, GPI p137, GPIAP1, GPIP137, M11S1, p137GPI, RNG105
Caprin1
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.