missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CAPRIN1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15112-1-AP-20UL
This item is not returnable.
View return policy
Description
GPIP137 may regulate the transport and translation of mRNAs of proteins involved in synaptic plasticity in neurons and cell proliferation and migration in multiple cell types. It is expressed in the cytoplasm.Specifications
| CAPRIN1 | |
| Polyclonal | |
| Unconjugated | |
| CAPRIN1 | |
| caprin 1, CAPRIN1, GPI anchored protein p137, GPI p137, GPIAP1, GPIP137, M11S1, p137GPI, RNG105 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 362173, 4076, 53872 | |
| -20°C | |
| Liquid |
| Western Blot, Immunofluorescence, Immunoprecipitation, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| 0.21 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q14444, Q5M9G3, Q60865 | |
| Caprin1 | |
| CAPRIN1 Fusion Protein Ag7099 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu