Learn More
CAPRIN1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 15112-1-AP-150UL
 View more versions of this product
Product Code. 16804144
  • £349.00 / 150µL
Description
GPIP137 may regulate the transport and translation of mRNAs of proteins involved in synaptic plasticity in neurons and cell proliferation and migration in multiple cell types. It is expressed in the cytoplasm.
Specifications
Specifications
CAPRIN1
Polyclonal
Unconjugated
CAPRIN1
caprin 1, CAPRIN1, GPI anchored protein p137, GPI p137, GPIAP1, GPIP137, M11S1, p137GPI, RNG105
Rabbit
Antigen Affinity Chromatography
RUO
362173, 4076, 53872
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.21 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q14444, Q5M9G3, Q60865
Caprin1
CAPRIN1 Fusion Protein Ag7099
150 μL
Primary
Human, Mouse, Rat