missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ C3a Receptor Recombinant Rabbit Monoclonal Antibody (6H9D5)

Rabbit Recombinant Monoclonal Antibody
£147.00 - £363.00
Specifications
| Antigen | C3a Receptor |
|---|---|
| Clone | 6H9D5 |
| Concentration | 2.69 mg/mL |
| Content And Storage | -20°C, Avoid Freeze/Thaw Cycles |
| Applications | ELISA, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30284402
|
Invitrogen™
MA558207 |
20 μL |
£147.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30283816
![]() |
Invitrogen™
MA558208 |
100 μL |
£363.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Immunogen sequence: MASFSAETNS TDLLSQPWNE PPVILSMVIL SLTFLLGLPG NGLVLWVAGL KMQRTVNTIW FLHLTLADLL CCLSLPFSLA HLALQGQWPY GRFLCKLIPS.
C3AR1 is a receptor for the chemotactic and inflammatory peptide anaphylatoxin C3a. The receptor stimulates chemotaxis, granule enzyme release and superoxide anion production. (Uniprot)Specifications
| C3a Receptor | |
| 2.69 mg/mL | |
| ELISA, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| O09047 | |
| 12267 | |
| Synthetic peptide. | |
| Antibody |
| 6H9D5 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Mouse | |
| C3ar1 | |
| anaphylatoxin C3a receptor; AZ3B; C3a anaphylatoxin chemotactic receptor; C3a receptor; C3AR; C3a-R; C3AR1; C3r1; complement component 3 receptor 1; complement component 3a receptor 1; HNFAG09 | |
| C3ar1 | |
| Primary | |
| Protein A |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title