missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ C3a Receptor Recombinant Rabbit Monoclonal Antibody (6H9D5)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA558207
This item is not returnable.
View return policy
Description
Immunogen sequence: MASFSAETNS TDLLSQPWNE PPVILSMVIL SLTFLLGLPG NGLVLWVAGL KMQRTVNTIW FLHLTLADLL CCLSLPFSLA HLALQGQWPY GRFLCKLIPS.
C3AR1 is a receptor for the chemotactic and inflammatory peptide anaphylatoxin C3a. The receptor stimulates chemotaxis, granule enzyme release and superoxide anion production. (Uniprot).
Specifications
| C3a Receptor | |
| Recombinant Monoclonal | |
| 2.69 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| O09047 | |
| C3ar1 | |
| Synthetic peptide. | |
| 20 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
| ELISA, Immunohistochemistry (Paraffin) | |
| 6H9D5 | |
| Unconjugated | |
| C3ar1 | |
| anaphylatoxin C3a receptor; AZ3B; C3a anaphylatoxin chemotactic receptor; C3a receptor; C3AR; C3a-R; C3AR1; C3r1; complement component 3 receptor 1; complement component 3a receptor 1; HNFAG09 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 12267 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction