missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ C3a Receptor Recombinant Rabbit Monoclonal Antibody (6H9D5)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA558208

 View more versions of this product

Product Code. 30283816

  • £363.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: MASFSAETNS TDLLSQPWNE PPVILSMVIL SLTFLLGLPG NGLVLWVAGL KMQRTVNTIW FLHLTLADLL CCLSLPFSLA HLALQGQWPY GRFLCKLIPS.

C3AR1 is a receptor for the chemotactic and inflammatory peptide anaphylatoxin C3a. The receptor stimulates chemotaxis, granule enzyme release and superoxide anion production. (Uniprot).
TRUSTED_SUSTAINABILITY
Specifications

Specifications

C3a Receptor
Recombinant Monoclonal
2.69 mg/mL
PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3
O09047
C3ar1
Synthetic peptide.
100 μL
Primary
Mouse
Antibody
IgG
ELISA, Immunohistochemistry (Paraffin)
6H9D5
Unconjugated
C3ar1
anaphylatoxin C3a receptor; AZ3B; C3a anaphylatoxin chemotactic receptor; C3a receptor; C3AR; C3a-R; C3AR1; C3r1; complement component 3 receptor 1; complement component 3a receptor 1; HNFAG09
Rabbit
Protein A
RUO
12267
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.