missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ C20orf3 Monoclonal Antibody (24GB6435)

Mouse Monoclonal Antibody
£172.00 - £488.00
Specifications
| Antigen | C20orf3 |
|---|---|
| Clone | 24GB6435 |
| Content And Storage | -20°C |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30276648
![]() |
Invitrogen™
MA557339 |
100 μL |
£488.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30275405
|
Invitrogen™
MA557340 |
20 μL |
£172.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
C20orf3 Monoclonal Antibody for Western Blot
APMAP (Adipocyte Plasma Membrane Associated Protein) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene include arylesterase activity and strictosidine synthase activity. Exhibits strong arylesterase activity with beta-naphthyl acetate and phenyl acetate. May play a role in adipocyte differentiation.Specifications
| C20orf3 | |
| -20°C | |
| Monoclonal | |
| Liquid | |
| IgG1 | |
| Human | |
| APMAP | |
| Adipocyte plasma membrane-associated protein; APMAP; C20orf3 | |
| APMAP | |
| Primary | |
| Affinity chromatography |
| 24GB6435 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9HDC9 | |
| 57136 | |
| Recombinant protein of human C20orf3 (aa 1-416). | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title