missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ C20orf3 Monoclonal Antibody (24GB6435)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA557339
This item is not returnable.
View return policy
Description
C20orf3 Monoclonal Antibody for Western Blot
APMAP (Adipocyte Plasma Membrane Associated Protein) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene include arylesterase activity and strictosidine synthase activity. Exhibits strong arylesterase activity with beta-naphthyl acetate and phenyl acetate. May play a role in adipocyte differentiation.
Specifications
| C20orf3 | |
| Monoclonal | |
| Unconjugated | |
| APMAP | |
| Adipocyte plasma membrane-associated protein; APMAP; C20orf3 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 57136 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB6435 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9HDC9 | |
| APMAP | |
| Recombinant protein of human C20orf3 (aa 1-416). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction