Learn More
Invitrogen™ C20orf3 Monoclonal Antibody (24GB6435)
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA557340
 View more versions of this product
Product Code. 30275405
  • £172.00 / 20µL
Description
C20orf3 Monoclonal Antibody for Western Blot
APMAP (Adipocyte Plasma Membrane Associated Protein) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene include arylesterase activity and strictosidine synthase activity. Exhibits strong arylesterase activity with beta-naphthyl acetate and phenyl acetate. May play a role in adipocyte differentiation.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
C20orf3
Monoclonal
Unconjugated
APMAP
Adipocyte plasma membrane-associated protein; APMAP; C20orf3
Mouse
Affinity chromatography
RUO
57136
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
24GB6435
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q9HDC9
APMAP
Recombinant protein of human C20orf3 (aa 1-416).
20 μL
Primary
Human
Antibody
IgG1