missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer solution pH 4, Phthalate buffer, traceable to NIST, colored red, Thermo Scientific™
£24.10 - £59.40
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10473053
|
Thermo Scientific Acros
383840010 |
1 L | Plastic bottle |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
10259243
|
Thermo Scientific Acros
383840025 |
2.5 L | Plastic bottle |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
pH buffer