Learn More
Buffer solution pH 4, Phthalate buffer, traceable to NIST, colored red, Thermo Scientific™
Brand: Thermo Scientific Acros 383840025
Code : 29
Additional Details : CAS Number : 877-24-7 Weight : 2.55000kg
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
pH bufferSpecifications
Buffer solution pH 4 | |
877-24-7,7732-18-5,7487-94-7,3244-88-0 | |
1.0000g/mL | |
Liquid | |
MFCD00217672 | |
1 | |
14, 7612 |
Phthalate buffer, traceable to NIST | |
Red | |
3.98 to 4.02 (20°C) | |
2.5 L | |
Plastic bottle | |
Traceable to NIST | |
Solubility in water: miscible. |