missing translation for 'onlineSavingsMsg'
Learn More

Buffer solution pH 4, Phthalate buffer, traceable to NIST, colored red, Thermo Scientific™

Brand:  Thermo Scientific Acros 383840010

Code : 29

Additional Details : CAS Number : 877-24-7 Weight : 1.05000kg

 View more versions of this product

Product Code. 10473053

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

pH buffer
Specifications

Specifications

Buffer solution pH 4
877-24-7,7732-18-5,7487-94-7,3244-88-0
1.0000g/mL
Liquid
MFCD00217672
1
14, 7612
Phthalate buffer, traceable to NIST
Red
3.98 to 4.02 (20°C)
1 L
Plastic bottle
Traceable to NIST
Solubility in water: miscible.
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Special Offers

Special Offers