missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ BS3 (bis(sulfosuccinimidyl)suberate)

Product Code. 11841245 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 mg
8 x 2 mg
10 x 2 mg
1 g
Unit Size:
1g
50mg
Pack of 10
Pack of 8
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11841245 8 x 2 mg Pack of 8
16091992 10 x 2 mg Pack of 10
10249963 50 mg 50mg
10075843 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11841245 Supplier Thermo Scientific™ Supplier No. 21585

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Crosslink amines to amines with BS3; composed of sulfo-NHS-ester groups separated by an 11.4A, water-soluble spacer arm that is useful for cell-surface protein crosslinking.

Thermo Scientific™ Pierce BS3 (Sulfo-DSS) is bis(sulfosuccinimidyl)suberate, an amine-to-amine crosslinker that is homobifunctional, water-soluble, non-cleavable and membrane impermeable.

Highlights:

  • Crosslink cell-surface proteins before cell lysis and immunoprecipitation
  • Identify receptor-ligand interactions by chemical crosslinking
  • “Fix” protein interactions to allow identification of weak or transient protein interactions
  • Identify near-neighbor protein interactions
  • Protein crosslinking to create bioconjugates via single-step reactions
  • Immobilize proteins onto amine-coated surfaces
TRUSTED_SUSTAINABILITY

Specifications

Chemical Reactivity Amine-to-Amine
Cleavable No
Formulation Bis(sulfosuccinimidyl) suberate; solid
Membrane Permeable No
Molecular Weight (g/mol) 572.43
PEGylated No
Sufficient For 28mL of typical 1X (1mM) reaction solution (total), or 8 separate uses
Spacer Arm Length 11.4Å
Quantity 8 x 2 mg
Solubility Water
Product Type Crosslinker
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.