Learn More
VEE GEE Brix Hydrometer
The Brix Hydrometer is calibrated to show the percentage of sucrose by weight at 20°C. The Brix scale is based on the following: 1°Brix = 1% sucrose by weight at the specified temperature (20°C).
£34.70
Product Code | Brand | Brix Range | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Brix Range | Price | Quantity & Availability | |||||
17896613
|
VEE GEE
6601-15 |
15 to 25° |
£34.70
Each |
Estimated Shipment: 26-06-2024
Log in to see stock. |
|||||
17866583
|
VEE GEE
6601-5 |
39 to 51° |
£34.70
Each |
Estimated Shipment: 26-06-2024
Log in to see stock. |
|||||
17826573
|
VEE GEE
6601-10 |
0 to 35° |
£34.70
Each |
Estimated Shipment: 26-06-2024
Log in to see stock. |
|||||
17896543
|
VEE GEE
6601-3 |
19 to 31° |
£34.70
Each |
Estimated Shipment: 26-06-2024
Log in to see stock. |
|||||
17836573
|
VEE GEE
6601-1 |
0 to 12° |
£34.70
Each |
Estimated Shipment: 26-06-2024
Log in to see stock. |
|||||
17856483
|
VEE GEE
6601-2 |
9 to 21° |
£34.70
Each |
Estimated Shipment: 26-06-2024
Log in to see stock. |
|||||
17876483
|
VEE GEE
6601-14 |
5 to 15° |
£34.70
Each |
Estimated Shipment: 26-06-2024
Log in to see stock. |
|||||
17846493
|
VEE GEE
6601-4 |
29 to 41° |
£34.70
Each |
Estimated Shipment: 26-06-2024
Log in to see stock. |
|||||
17836483
|
VEE GEE
6601-11 |
35 to 70° |
£34.70
Each |
Estimated Shipment: 26-06-2024
Log in to see stock. |
|||||
17866483
|
VEE GEE
6601-13 |
-5 to +5° |
£34.70
Each |
Estimated Shipment: 26-06-2024
Log in to see stock. |
|||||
Description
- Approx. Length: 330 mm
Specifications
Brix Hydrometer | |
33 cm |