missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VEE GEE Brix Hydrometer
The Brix Hydrometer is calibrated to show the percentage of sucrose by weight at 20°C. The Brix scale is based on the following: 1°Brix = 1% sucrose by weight at the specified temperature (20°C).
Brand: VEE GEE 6601-3
Additional Details : Weight : 0.11000kg
Description
- Approx. Length: 330 mm
Specifications
Brix Hydrometer | |
33 cm | |
20°C |
0.1° | |
19 to 31° |