Learn More
Fisherbrand™ Borosilicate Glass Squat Form Heavy Duty Beakers
Product Code. 15476123 Shop All Fisherbrand Products
Click to view available options
Capacity (Metric):
150 mL
250 mL
400 mL
600 mL
1000 mL
2000 mL
Unit Size:
Pack of 10
Pack of 4
Product Code. 15476123
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Feature heavy wall construction for increased mechanical strength. Fisherbrand™ Borosilicate Glass Squat Form Heavy Duty Beakers are made of full 3.3 expansion borosilicate glass and feature an 'easy pour' spout.
  • Heavy wall construction for increased mechanical strength
  • Much more robust than standard beakers
  • Full 3.3 expansion borosilicate glass
  • Reinforced rim with 'easy pour' pouring spout
  • Graduated in white ceramic with marking area
  • Uniform wall thickness for optimum optical clarity
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 250 mL
Material Borosilicate Glass
Includes Spout
Quantity 10
Type Beaker
Graduated Yes
Scale White Ceramic Scale
Style Squat Form