missing translation for 'onlineSavingsMsg'
Learn More

Robu™ Borosilicate Glass Micro-Filter Funnels

Brand:  Robu™ 28044

Additional Details : Weight : 0.06460kg

 View more versions of this product

Product Code. 16893076

  • £137.00 / Pack of 5
Estimated Shipment: 17-05-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

Micro filter funnel
6mm
55 mm
Unground joint
4 mL
Borosilicate Glass
Highly resistant to most chemicals such as solvents, acids and alkalis
4
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers