Learn More
Robu™ Borosilicate Glass Micro-Filter Funnels
Brand:  Robu™ 27083
Additional Details : Weight : 0.03990kg
 View more versions of this product
Product Code. 16803076
  • £103.00 / Pack of 5
Estimated Shipment: 16-05-2024
to see stock.
Add to basket
Specifications
Micro filter funnel
0.8 mL
Borosilicate Glass
Highly resistant to most chemicals such as solvents, acids and alkalis
3
10mm
6mm
20 mm
10mm