Learn More
Robu™ Borosilicate Glass Micro-Filter Funnels
Brand: Robu™ 27084
Additional Details : Weight : 0.10000kg
Especificaciones
Micro filter funnel | |
0.8 mL | |
Borosilicate Glass | |
Highly resistant to most chemicals such as solvents, acids and alkalis | |
4 |
10mm | |
6mm | |
20 mm | |
10mm |