missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ BMP-7 Recombinant Rabbit Monoclonal Antibody (23GB4845), Invitrogen™
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

£162.00 - £387.00

Specifications

Antigen BMP-7
Clone 23GB4845
Applications Flow Cytometry, Western Blot
Classification Recombinant Monoclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30087489
GREENER_CHOICE
View Documents
Invitrogen™
MA552760
100 μL
£387.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30088512
View Documents
Invitrogen™
MA552761
20 μL
£162.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

BMP-7 Recombinant Monoclonal antibody specifically detects BMP-7 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Western Blot
Specifications

Specifications

BMP-7
Flow Cytometry, Western Blot
Unconjugated
Rabbit
Human, Mouse, Rat
P18075, P23359
12162, 655, 85272
IgG
Affinity chromatography
Antibody
23GB4845
Recombinant Monoclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
BMP; BMP7; BMP-7; bone moorphogenic protein-7; Bone morphogenetic protein; bone morphogenetic protein 7; bone morphogenetic protein 7 (osteogenic protein 1); bone morphogenetic protein 7 preproprotein; bone morphogenic protein 7; Eptotermin alfa; OP1; OP-1; Osteogenic protein 1; RP11-560A15.5
A synthesized peptide derived from human BMP7 (300-350AA).
Primary
-20°C
Bmp7
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.