missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BMP-7 Recombinant Rabbit Monoclonal Antibody (23GB4845), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA552761
This item is not returnable.
View return policy
Description
The bone morphogenetic proteins (BMPs) are a family of secreted signaling molecules that can induce ectopic bone growth. Many BMPs are part of the transforming growth factor-beta (TGFB) superfamily. BMPs were originally identified by an ability of demineralized bone extract to induce endochondral osteogenesis in vivo in an extra skeletal site. Based on its expression early in embryogenesis, BMP7 has a proposed role in early development. In addition, the fact that BMP7 is closely related to BMP5 and BMP6 has lead to speculation of possible bone inductive activity.
Specifications
| BMP-7 | |
| Recombinant Monoclonal | |
| 1.55 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P18075, P23359 | |
| Bmp7 | |
| A synthesized peptide derived from human BMP7 (300-350AA). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot | |
| 23GB4845 | |
| Unconjugated | |
| Bmp7 | |
| BMP; BMP7; BMP-7; bone moorphogenic protein-7; Bone morphogenetic protein; bone morphogenetic protein 7; bone morphogenetic protein 7 (osteogenic protein 1); bone morphogenetic protein 7 preproprotein; bone morphogenic protein 7; Eptotermin alfa; OP1; OP-1; Osteogenic protein 1; RP11-560A15.5 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 12162, 655, 85272 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction