missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ BMP-7 Recombinant Rabbit Monoclonal Antibody (23GB4845), Invitrogen™
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA552760

 View more versions of this product

Product Code. 30087489

  • £387.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The bone morphogenetic proteins (BMPs) are a family of secreted signaling molecules that can induce ectopic bone growth. Many BMPs are part of the transforming growth factor-beta (TGFB) superfamily. BMPs were originally identified by an ability of demineralized bone extract to induce endochondral osteogenesis in vivo in an extra skeletal site. Based on its expression early in embryogenesis, BMP7 has a proposed role in early development. In addition, the fact that BMP7 is closely related to BMP5 and BMP6 has lead to speculation of possible bone inductive activity.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

BMP-7
Recombinant Monoclonal
1.55 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P18075, P23359
Bmp7
A synthesized peptide derived from human BMP7 (300-350AA).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Flow Cytometry, Western Blot
23GB4845
Unconjugated
Bmp7
BMP; BMP7; BMP-7; bone moorphogenic protein-7; Bone morphogenetic protein; bone morphogenetic protein 7; bone morphogenetic protein 7 (osteogenic protein 1); bone morphogenetic protein 7 preproprotein; bone morphogenic protein 7; Eptotermin alfa; OP1; OP-1; Osteogenic protein 1; RP11-560A15.5
Rabbit
Affinity chromatography
RUO
12162, 655, 85272
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.