missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Biotin-conjugated t-Plasminogen activator/tPA Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
£119.00 - £295.00
Specifications
| Antigen | t-Plasminogen activator/tPA |
|---|---|
| Concentration | 1000 μg/mL |
| Applications | Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | Biotin |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30236822
|
Proteintech
BIOTIN-10147-100UL |
100 μL |
£350.00 £295.00 / 100µL Save £55.00 15% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30236813
|
Proteintech
BIOTIN-10147-20UL |
20 μL |
£130.00 £119.00 / 20µL Save £11.00 8% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The t-Plasminogen activator/tPA antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human t-Plasminogen activator/tPA. This antibody recognizes human, mouse, rat antigen. The t-Plasminogen activator/tPA antibody has been validated for the following applications: IHC analysis. This antibody is biotin.Specifications
| t-Plasminogen activator/tPA | |
| Immunohistochemistry | |
| Biotin | |
| Rabbit | |
| Human, Mouse, Rat | |
| DKFZp686I03148; T-PA; TPA | |
| IgG | |
| Antigen affinity purification | |
| Conjugated Antibody |
| 1000 μg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| 5327 | |
| Recombinant Protein | |
| Primary | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title