missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Biotin-conjugated t-Plasminogen activator/tPA Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech BIOTIN-10147-20UL
This item is not returnable.
View return policy
Description
The t-Plasminogen activator/tPA antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human t-Plasminogen activator/tPA. This antibody recognizes human, mouse, rat antigen. The t-Plasminogen activator/tPA antibody has been validated for the following applications: IHC analysis. This antibody is biotin.Specifications
| t-Plasminogen activator/tPA | |
| Polyclonal | |
| Biotin | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 5327 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Conjugated Antibody | |
| IgG |
| Immunohistochemistry | |
| 1000 μg/mL | |
| DKFZp686I03148; T-PA; TPA | |
| Recombinant Protein | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| 32-35 kDa, 65 kDa | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction