missing translation for 'onlineSavingsMsg'
Learn More

Biotin-conjugated t-Plasminogen activator/tPA Rabbit Polyclonal Antibody, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech BIOTIN-10147-100UL

 View more versions of this product

Product Code. 30236822

  • ONLINE EXCLUSIVE PRICE
    £350.00
    £295.00 / 100µL
    Save £55.00
    15% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The t-Plasminogen activator/tPA antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human t-Plasminogen activator/tPA. This antibody recognizes human, mouse, rat antigen. The t-Plasminogen activator/tPA antibody has been validated for the following applications: IHC analysis. This antibody is biotin.
Specifications

Specifications

t-Plasminogen activator/tPA
Polyclonal
Biotin
Rabbit
Antigen affinity purification
RUO
5327
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.
Conjugated Antibody
IgG
Immunohistochemistry
1000 μg/mL
DKFZp686I03148; T-PA; TPA
Recombinant Protein
100 μL
Primary
Human, Mouse, Rat
32-35 kDa, 65 kDa
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.