missing translation for 'onlineSavingsMsg'
Learn More

BID Mouse anti-Human, HRP, Clone: 2A2E5, Proteintech

Mouse Monoclonal Antibody

£135.00 - £375.00

Specifications

Antigen BID
Clone 2A2E5
Applications Western Blot
Classification Monoclonal
Conjugate HRP
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17741489
View Documents
Proteintech
HRP-60301-20UL
20 μL
£135.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16841246
View Documents
Proteintech
HRP-60301-100UL
100 μL
£375.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The BID antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human BID. This antibody recognizes human antigen. The BID antibody has been validated for the following applications: WB analysis. This antibody is hrp.

Specifications

Specifications

BID
Western Blot
HRP
Human
IgG1
2A2E5
Monoclonal
Mouse
637
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.