Learn More
BID Mouse anti-Human, HRP, Clone: 2A2E5, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech HRP-60301-20UL
This item is not returnable.
View return policy
Description
The BID antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human BID. This antibody recognizes human antigen. The BID antibody has been validated for the following applications: WB analysis. This antibody is hrp.
Specifications
| BID | |
| Monoclonal | |
| 1000 ug/mL | |
| Mouse | |
| Primary | |
| Human |
| Western Blot | |
| 2A2E5 | |
| HRP | |
| 20 μL | |
| 637 | |
| IgG1 |