Learn More
BID Mouse anti-Human, HRP, Clone: 2A2E5, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech HRP-60301-100UL
 View more versions of this product
Product Code. 16841246
  • £375.00 / 100µL
Description
The BID antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human BID. This antibody recognizes human antigen. The BID antibody has been validated for the following applications: WB analysis. This antibody is hrp.
Specifications
Specifications
BID
Monoclonal
1000 μg/mL
Mouse
Primary
Human
Western Blot
2A2E5
HRP
100 μL
637
IgG1