missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta-Actin Antibody (CL3868), Novus Biologicals™
Mouse Monoclonal Antibody
£240.00 - £442.00
Specifications
| Antigen | beta-Actin |
|---|---|
| Clone | CL3868 |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18337060
|
Novus Biologicals
NBP2-76481 |
100 μL |
£442.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18653721
|
Novus Biologicals
NBP2-76481-25ul |
25 μL |
£240.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
beta-Actin Monoclonal specifically detects beta-Actin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| beta-Actin | |
| Monoclonal | |
| Mouse | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% sodium azide | |
| 60 | |
| This beta-Actin Antibody (CL3868) was developed against a recombinant protein corresponding to amino acids: PKANREKMTQIMFETFNTPAMYVAIQ | |
| Primary | |
| 42 kDa |
| CL3868 | |
| Unconjugated | |
| Human, Mouse, Rat | |
| ACTB, actin, beta, actin, cytoplasmic 1, B-Actin, Beta Actin, beta cytoskeletal actin, Beta-actin, BRWS1, I(2)-Actin, PS1TP5-binding protein 1, PS1TP5BP1 | |
| ACTB | |
| IgG1 | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title