missing translation for 'onlineSavingsMsg'
Learn More

beta-Actin Antibody (CL3868), Novus Biologicals™

Product Code. 18337060 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
Unit Size:
100µL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18337060 100 μL 100µL
18653721 25 μL 25µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18337060 Supplier Novus Biologicals Supplier No. NBP276481

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

beta-Actin Monoclonal specifically detects beta-Actin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen beta-Actin
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CL3868
Conjugate Unconjugated
Dilution Western Blot 1 ug/ml, Immunohistochemistry 1:5000 - 1:10000, Immunocytochemistry/Immunofluorescence 2-10 ug/ml, Immunohistochemistry-Paraffin 1:5000 - 1:10000
Formulation PBS, pH 7.2, containing 40% glycerol with 0.02% sodium azide
Gene Alias ACTB, actin, beta, actin, cytoplasmic 1, B-Actin, Beta Actin, beta cytoskeletal actin, Beta-actin, BRWS1, I(2)-Actin, PS1TP5-binding protein 1, PS1TP5BP1
Gene Symbols ACTB
Host Species Mouse
Immunogen This beta-Actin Antibody (CL3868) was developed against a recombinant protein corresponding to amino acids: PKANREKMTQIMFETFNTPAMYVAIQ
Molecular Weight of Antigen 42 kDa
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 60
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.