missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta-Actin Antibody (CL3868), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-76481-25ul
This item is not returnable.
View return policy
Description
beta-Actin Monoclonal specifically detects beta-Actin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| beta-Actin | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% sodium azide | |
| ACTB | |
| This beta-Actin Antibody (CL3868) was developed against a recombinant protein corresponding to amino acids: PKANREKMTQIMFETFNTPAMYVAIQ | |
| 25 μL | |
| 60 | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| CL3868 | |
| Western Blot 1 μg/mL, Immunohistochemistry 1:5000 - 1:10000, Immunocytochemistry/Immunofluorescence 2-10 μg/mL, Immunohistochemistry-Paraffin 1:5000 - 1:10000 | |
| ACTB, actin, beta, actin, cytoplasmic 1, B-Actin, Beta Actin, beta cytoskeletal actin, Beta-actin, BRWS1, I(2)-Actin, PS1TP5-binding protein 1, PS1TP5BP1 | |
| Mouse | |
| 42 kDa | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction