missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ SP Scienceware™ Write-On™ Colored Label Tapes
Custom manufactured from proprietary materials that resist moisture and most solvents
£11.86 - £22.88
Product Code | Brand | Color | Width (English) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Width (English) | Price | Quantity & Availability | ||||
12368797
|
Bel-Art™
134623010 |
Green | 1 in. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12388787
|
Bel-Art™
134620005 |
White | 0.5 in. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12348797
|
Bel-Art™
134623005 |
0.5 in. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
12328797
|
Bel-Art™
134622075 |
Yellow | 0.75 in. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12318797
|
Bel-Art™
134622005 |
0.5 in. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
12358797
|
Bel-Art™
134623075 |
Green | 0.75 in. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12338777
|
Bel-Art™
F134631005 |
0.5 in. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
12348777
|
Bel-Art™
F134631075 |
Red | 0.75 in. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12358777
|
Bel-Art™
F134631010 |
Red | 1 in. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12328787
|
Bel-Art™
F134634005 |
Blue | 0.5 in. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12338797
|
Bel-Art™
134622010 |
Yellow | 1 in. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12348787
|
Bel-Art™
134634010 |
Blue | 1 in. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12328777
|
Bel-Art™
F134635010 |
1 in. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
12398787
|
Bel-Art™
134620075 |
0.75 in. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
12378767
|
Bel-Art™
134630005 |
0.5 in. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
12388767
|
Bel-Art™
F134630075 |
White | 0.75 in. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
Colors correspond with familiar hazard color codes to highlight safety information written on the labels.
- Adhere to TFE fluoropolymer resin covered materials and any clean surface
- Can endure a maximum temperature of 247°C (492°F) for 30 minutes
- Samples stay labeled even after steam autoclaving at 121°C (250°F) or freezing down to -73°C (-99°F)
- No sticky residue lingers when tape is stripped off
- Marks made with pencil, ball-point pen or solvent ink marker survive most laboratory conditions