missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ SP Scienceware™ Write-On™ Colored Label Tapes
Custom manufactured from proprietary materials that resist moisture and most solvents
Brand: Bel-Art™ 134634010
Code : 03
Additional Details : Weight : 0.01000kg
Description
Colors correspond with familiar hazard color codes to highlight safety information written on the labels.
- Adhere to TFE fluoropolymer resin covered materials and any clean surface
- Can endure a maximum temperature of 247°C (492°F) for 30 minutes
- Samples stay labeled even after steam autoclaving at 121°C (250°F) or freezing down to -73°C (-99°F)
- No sticky residue lingers when tape is stripped off
- Marks made with pencil, ball-point pen or solvent ink marker survive most laboratory conditions
Specifications
Blue | |
Tape | |
1 in. |