Learn More
Axygen™ Automation Tips for PerkinElmer™, 96-Well
Ideal for Janus™ MiniTrak, PlateTrak™ and Evolution P3. Axygen™ Automation Tips for PerkinElmer™, 96-Well are constructed of clear polypropylene. Available in sterile/non-sterile, filtered/non-filtered options, from 20 to 200 μL.
Brand:  Axygen™ PK-200-R
427.58 GBP valid until 2024-12-31
Use promo code "23272" to get your promotional price.
Code : 68
Additional Details : Weight : 9.98000kg
 View more versions of this product
Product Code. 12752896
  • £449.00 / Pack of 4800
Estimated Shipment: 26-09-2024
to see stock.
Add to basket
Description
Description
Assembled via automation under strict process controls to ensure consistent performance and quality
  • Robotic tips use no mold-releasing agents
  • Nucleic acid, nuclease and endotoxin-safe for complete protection against contamination
  • Tips are certified RNase-/DNase-free and nonpyrogenic
  • Compatible with PerkinElmer™ MDT-96 P235, MDT-96 P50 and MDT-384 30μL head for:
    • Janus™
    • Evolution P3
    • Minitrak
    • PlateTrak™
Multiple options
  • Presterilized and non-sterile tips
  • With or without aerosol-resistant polyethylene filters
  • Wide bore series for working with viscous materials, genomic DNA or delicate cell cultures
  • Available in a range of volumes: 20, 30, 50, 180 and 200μL
Specifications
Specifications
200 μL
Non-sterile
Green
DNase, Rnase Free
Rack
SBS Rack
PerkinElmer Janus, PerkinElmer Evolution P3, PerkinElmer MiniTrak, PerkinElmer PlateTrak, PerkinElmer MDT-96 P235, PerkinElmer MDT-96 P50, PerkinElmer MDT-384 30μL
Non-Filtered
Clear
ANSI
Non-pyrogenic, used for stringent specifications under strict process controls, nucleic acid, nuclease and endotoxin-safe for complete protection against contamination
Automation, Round
50 Racks/Cs.
Polypropylene